missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7124763
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C10555G 5 g
C105525G 25 g
2 options

Catalog No. C10555G

Supplier: TCI America C10555G

Only null left

Chemical Identifiers

CAS 1878-65-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004332
InChI Key WFPMUFXQDKMVCO-UHFFFAOYSA-N
Synonym 3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
PubChem CID 15879
IUPAC Name 2-(3-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Cl)=C1

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Chlorophenylacetic Acid