accessibility menu, dialog, popup

UserName

CAS RN 1878-65-5

IUPAC Name: 2-(3-chlorophenyl)acetic acid
Synonyms: M-chlorophenylacetic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: WFPMUFXQDKMVCO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)