missing translation for 'onlineSavingsMsg'
Learn More

Acide 3-chlorophénylacétique 98,0+%, TCI America™

Numéro de catalogue. C10555G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C10555G 5 g
C105525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C10555G Fournisseur TCI America Code fournisseur C10555G

Identifiants chimiques

CAS 1878-65-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004332
InChI Key WFPMUFXQDKMVCO-UHFFFAOYSA-N
Synonym 3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
PubChem CID 15879
IUPAC Name Acide 2-(3-chlorophényl)acétique
SMILES OC(=O)CC1=CC=CC(Cl)=C1

Spécifications

Melting Point 81°C
Color Blanc-jaune
Quantity 5 g
Physical Form Poudre de cristal à 20 °C
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 3-Chlorophenylacetic Acid