missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroperoxybenzoic Acid, 70-75%, balance 3-Chlorobenzoic Acid and water

Catalog No. AC255790010
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255790010 1 kg Plastic bottle
AC255790250 25 g Plastic bottle
AC255791000 100 g Plastic bottle
AC255795000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC255790010 Supplier Thermo Scientific Chemicals Supplier No. 255790010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5 | CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Specifications

Chemical Name or Material 3-Chloroperoxybenzoic acid
Name Note 70 - 75%
CAS 937-14-4, 535-80-8, 7732-18-5
CAS Min % 70.0, 25.0, 25.0
CAS Max % 75.0, 30.0, 30.0
Melting Point 92.0°C to 94.0°C
Color White
Density 0.5600g/mL
Assay Percent Range di-m-Chlorobenzoyl peroxide: < 0.1% Typical (HPLC)
Packaging Plastic bottle
Molecular Formula C7H5ClO3
Linear Formula ClC6H4CO3H
Quantity 1 kg
MDL Number MFCD00002127
Beilstein 09,IV,972
Fieser 01,135; 02,68; 03,49; 04,85; 05,120; 06,110; 07,62; 08,97; 09,108; 10,92; 11,122; 12,118; 13,76; 15,86; 16,80; 17,76
Merck Index 15,2154
Specific Gravity 0.56
Synonym 3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in koh, soluble in chloroform and acetone
InChI Key NHQDETIJWKXCTC-UHFFFAOYSA-N
SMILES OOC(=O)C1=CC=CC(Cl)=C1
IUPAC Name 3-chlorobenzenecarboperoxoic acid
Molecular Weight (g/mol) 172.56
PubChem CID 70297
ChEBI CHEBI:52091
Formula Weight 172.57
Percent Purity 70-75%
Physical Form Moist Powder
Show More Show Less

RUO – Research Use Only