missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)benzoyl Chloride 93.0+%, TCI America™
Supplier: TCI America C231425G
Specifications
| 3-(Chloromethyl)benzoyl Chloride | |
| 4°C | |
| 150°C | |
| MFCD00000682 | |
| 3265 | |
| YCAIYRWHKSJKEB-UHFFFAOYSA-N | |
| 3-(chloromethyl)benzoyl chloride | |
| 2733324 | |
| ≥93.0% (GC) |
| 63024-77-1 | |
| Yellow | |
| C8H6Cl2O | |
| 25 g | |
| 3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n | |
| C1=CC(=CC(=C1)CCl)C(=O)Cl | |
| 189.035 | |
| 189.04 | |
| Liquid |
Chemical Identifiers
| 63024-77-1 | |
| 189.035 | |
| YCAIYRWHKSJKEB-UHFFFAOYSA-N | |
| 2733324 | |
| C1=CC(=CC(=C1)CCl)C(=O)Cl |
| C8H6Cl2O | |
| MFCD00000682 | |
| 3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n | |
| 3-(chloromethyl)benzoyl chloride |
Safety and Handling
TSCA : No