missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)benzoyl Chloride 93.0+%, TCI America™

Catalog No. C23145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23145G 5 g
C231425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23145G Supplier TCI America Supplier No. C23145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63024-77-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00000682
InChI Key YCAIYRWHKSJKEB-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n
PubChem CID 2733324
IUPAC Name 3-(chloromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)CCl)C(=O)Cl

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 150°C
Quantity 5 g
UN Number 3265
Formula Weight 189.04
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Chloromethyl)benzoyl Chloride