missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)benzoyl chloride, 98%

Catalog No. p-3735979
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314430050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314430050 Supplier Thermo Scientific Chemicals Supplier No. 314430050

CAS: 63024-77-1 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63024-77-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000682
InChI Key YCAIYRWHKSJKEB-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoyl chloride, 3-chloromethyl benzoylchloride, 3-chloromethyl-benzoyl chloride, benzoyl chloride, 3-chloromethyl, pubchem16448, 3-chlormethylbenzoylchloride, 3-chloromethylbenzoylchloride, acmc-1b65n
PubChem CID 2733324
IUPAC Name 3-(chloromethyl)benzoyl chloride
SMILES C1=CC(=CC(=C1)CCl)C(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3300g/mL
Boiling Point 149°C to 150°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C6H4COCl
Refractive Index 1.5738 to 1.5748
Quantity 5 g
Specific Gravity 1.33
Formula Weight 189.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(chloromethyl)benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 263-799-7

RUO – Research Use Only