missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1,2,4-triazolin-5-one 98.0+%, TCI America™

Catalog No. C30941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30941G 1 g
C30945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30941G Supplier TCI America Supplier No. C30941G

Chemical Identifiers

CAS 252742-72-6
Molecular Formula C3H4ClN3O
Molecular Weight (g/mol) 133.535
MDL Number MFCD07776630
InChI Key ZLRBJVJEQXBAAI-UHFFFAOYSA-N
Synonym 3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one
PubChem CID 10129979
IUPAC Name 5-(chloromethyl)-1,2-dihydro-1,2,4-triazol-3-one
SMILES C(C1=NC(=O)NN1)Cl

Specifications

Melting Point 199°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 133.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Chloromethyl)-1,2,4-triazolin-5-one