accessibility menu, dialog, popup

UserName

NOHNHNCl

CAS RN 252742-72-6

IUPAC Name: 5-(chloromethyl)-2,3-dihydro-1H-1,2,4-triazol-3-one
Synonyms: 5-(chloromethyl)-1,2-dihydro-1,2,4-triazol-3-one
Molecular Weight (g/mol): 133.54
Molecular Formula: C3H4ClN3O
InChi Key: ZLRBJVJEQXBAAI-UHFFFAOYSA-N
SMILES: ClCC1=NC(=O)NN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results