missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorocoumarin 98.0+%, TCI America™

Catalog No. C22971G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22971G Supplier TCI America Supplier No. C22971G
Only null left

Chemical Identifiers

CAS 92-45-5
Molecular Formula C9H5ClO2
Molecular Weight (g/mol) 180.59
MDL Number MFCD00024073
InChI Key CKCOPMSBJBNBCQ-UHFFFAOYSA-N
Synonym 3-chlorocoumarin, coumarin, 3-chloro, 2h-1-benzopyran-2-one, 3-chloro, 3-chloro-2h-chromen-2-one, chlorocoumarin, 3-chloro-2h-1-benzopyran-2-one, acmc-1ciat, 2h-1-benzopyran-2-one,3-chloro
PubChem CID 96133
IUPAC Name 3-chloro-2H-chromen-2-one
SMILES ClC1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 123°C
Quantity 1 g
Formula Weight 180.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorocoumarin