missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobutyric Acid 98.0+%, TCI America™

Catalog No. C014725ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C014725ML 25 mL
C014710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C014725ML Supplier TCI America Supplier No. C014725ML

Chemical Identifiers

CAS 1951-12-8
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00020496
InChI Key XEEMVPPCXNTVNP-UHFFFAOYNA-N
PubChem CID 16048
IUPAC Name 3-chlorobutanoic acid
SMILES CC(CC(=O)O)Cl

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 mL
UN Number 3265
Formula Weight 122.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chlorobutyric Acid
RTECSNumber ES8137500
TSCA Yes