accessibility menu, dialog, popup

UserName

CH3ClOHO

CAS RN 1951-12-8

IUPAC Name: 3-chlorobutanoic acid
Synonyms: 3-chlorobutanoic acid
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: XEEMVPPCXNTVNP-UHFFFAOYNA-N
SMILES: CC(Cl)CC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results