missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylideneacetone, 98%

Catalog No. p-7080694
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAA1033206 5 g
AAA1033214 25 g
2 options

Catalog No. AAA1033206

Supplier: Thermo Scientific Chemicals A1033206

Only null left

CAS: 20766-36-3 | C10H9ClO | 180.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20766-36-3
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00052851
InChI Key VWEPXSRBBXPYSM-AATRIKPKSA-N
Synonym 3e-4-3-chlorophenyl but-3-en-2-one, e-4-3-chloro-phenyl-but-3-en-2-one, 4-3-chlorophenyl but-3-en-2-one, e-4-3-chlorophenyl-but-3-en-2-one, 3-chlorobenzylideneacetone, 3-chloro-benzalacetone, 3-chlorostyrylmethyl ketone, 3-chlorobenzylidene acetone, chembl73680, e-4-3-chlorophenyl but-3-en-2-one
PubChem CID 5373975
IUPAC Name (E)-4-(3-chlorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=CC=C1)Cl

Specifications

Boiling Point 123°C to 125°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 180.64
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzylideneacetone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only