missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylamine, 98%

Catalog No. p-85356
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297570050 5 g Glass bottle
AC297570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297570050 Supplier Thermo Scientific Chemicals Supplier No. 297570050

CAS: 4152-90-3 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4152-90-3
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00040752
InChI Key BJFPYGGTDAYECS-UHFFFAOYSA-N
Synonym 3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
PubChem CID 77802
IUPAC Name (3-chlorophenyl)methanamine
SMILES C1=CC(=CC(=C1)Cl)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 110.0°C to 112.0°C (17.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5575 to 1.5605
Linear Formula ClC6H4CH2NH2
Quantity 5 g
Specific Gravity 1.15
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 223-985-0
TSCA TSCA

RUO – Research Use Only