missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl Cyanide 98.0+%, TCI America™

Catalog No. C109125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C109125G Supplier TCI America Supplier No. C109125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1529-41-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00001908
InChI Key GTIKLPYCSAMPNG-UHFFFAOYSA-N
Synonym 3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
PubChem CID 73722
IUPAC Name 2-(3-chlorophenyl)acetonitrile
SMILES C1=CC(=CC(=C1)Cl)CC#N

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
UN Number 3276
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzyl Cyanide
EINECSNumber (3)-1784
RTECSNumber AL8240000
TSCA No