missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl Chloride 98.0+%, TCI America™

Catalog No. C058025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058025G 25 g
C0580500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C058025G Supplier TCI America Supplier No. C058025G

Chemical Identifiers

CAS 620-20-2
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000905
InChI Key DDGRAFHHXYIQQR-UHFFFAOYSA-N
Synonym 3-chlorobenzyl chloride, 1-chloro-3-chloromethyl benzene, alpha,3-dichlorotoluene, m-chlorobenzyl chloride, benzene, 1-chloro-3-chloromethyl, 3-chlorobenzylchloride, m-chlorbenzyl chloride, 1-chloro-3-chloromethyl-benzene, toluene, m,.alpha.-dichloro, m,.alpha.-dichlorotoluene
PubChem CID 12103
IUPAC Name 1-chloro-3-(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)Cl)CCl

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 2235
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzyl Chloride
EINECSNumber (3)-0091
TSCA Yes