missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl Bromide 95.0+%, TCI America™

Catalog No. C110525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C110525G Supplier TCI America Supplier No. C110525G

Chemical Identifiers

CAS 766-80-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00000597
InChI Key LZIYAIRGDHSVED-UHFFFAOYSA-N
Synonym 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide
PubChem CID 69838
IUPAC Name 1-(bromomethyl)-3-chlorobenzene
SMILES ClC1=CC=CC(CBr)=C1

Specifications

Color Yellow
Boiling Point 109°C
Quantity 25 g
UN Number 3265
Formula Weight 205.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzyl Bromide