missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl bromide, 97%

Catalog No. AAA1584418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1584418 50 g
AAA1584409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1584418 Supplier Thermo Scientific Chemicals Supplier No. A1584418

CAS: 766-80-3 | C7H6BrCl | 205.48 g/mol

3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorobenzyl bromide was used in the synthesis of symmetrical and unsymmetrical benzyl thioethers. It was used as starting reagent during the synthesis of 1-(3-chlorobenzyl)-2-(pyrrolidin-1-ylmethyl)-1H-benzimidazole dihydrochloride.

Solubility
Decomposes in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 766-80-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00000597
InChI Key LZIYAIRGDHSVED-UHFFFAOYSA-N
Synonym 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide
PubChem CID 69838
IUPAC Name 1-(bromomethyl)-3-chlorobenzene
SMILES ClC1=CC=CC(CBr)=C1

Specifications

Density 1.567
Boiling Point 108°C to 110°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.589
Quantity 50 g
UN Number UN3265
Beilstein 636504
Solubility Information Decomposes in water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-171-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only