missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoylacetonitrile, 98%

Catalog No. p-7021539
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1391303 1 g
AAA1391306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1391303 Supplier Thermo Scientific Chemicals Supplier No. A1391303
Only null left

CAS: 21667-62-9 | C9H6ClNO | 179.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21667-62-9
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.60
MDL Number MFCD00067891
InChI Key IUDFNNHFARLIPF-UHFFFAOYSA-N
Synonym 3-chlorobenzoylacetonitrile, 3-3-chlorophenyl-3-oxopropanenitrile, 3-chlorophenacylcyanide, 3-chlorobenzoyl acetonitrile, 3-3-chlorophenyl-3-oxo-propanenitrile, beta-ketonitrile 5b, pubchem12046, m-chlorophenacyl cyanide, acmc-1cqmb, maybridge1_004670
PubChem CID 140855
IUPAC Name 3-(3-chlorophenyl)-3-oxopropanenitrile
SMILES ClC1=CC=CC(=C1)C(=O)CC#N

Specifications

Melting Point 78°C to 83°C
Quantity 1 g
Beilstein 2414476
Formula Weight 179.61
Percent Purity 98%
Chemical Name or Material 3-Chlorobenzoylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only