accessibility menu, dialog, popup

UserName

CAS RN 21667-62-9

IUPAC Name: 3-(3-chlorophenyl)-3-oxopropanenitrile
Synonyms: 3-(3-chlorophenyl)-3-oxopropanenitrile
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: IUDFNNHFARLIPF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C(=O)CC#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)