missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. c013925g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
C013925G 25 g
1 options

Catalog No. C013925G

Supplier: TCI America C013925G

Only null left

Chemical Identifiers

CAS 618-46-2
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000671
InChI Key WHIHIKVIWVIIER-UHFFFAOYSA-N
Synonym m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
PubChem CID 69252
IUPAC Name 3-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 222°C
Quantity 25 g
UN Number 3265
Formula Weight 175.01
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzoyl Chloride
EINECSNumber (3)-0062&(3)-1409
TSCA Yes