missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoyl chloride, 99+%

Catalog No. AC108740050
Change view
Click to view available options
Quantity:
5 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108740050 5 mL Glass bottle
AC108740500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108740050 Supplier Thermo Scientific Chemicals Supplier No. 108740050

CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-46-2
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000671
InChI Key WHIHIKVIWVIIER-UHFFFAOYSA-N
Synonym m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
PubChem CID 69252
IUPAC Name 3-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 1.3600g/mL
Boiling Point 223.0°C to 225.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 0.03% max. Total other poly-halogenated acid halides (GC)
Packaging Glass bottle
Refractive Index 1.5680 to 1.5700
Linear Formula ClC6H4COCl
Quantity 5 mL
Beilstein 09, 338
Specific Gravity 1.36
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzoyl chloride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 210-552-6
TSCA TSCA

RUO – Research Use Only