missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoyl chloride, 97%

CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol

Supplier:  Thermo Scientific Chemicals A1350922

 View more versions of this product

Catalog No. AAA1350922


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chlorobenzoyl chloride
1.368
117°C (242°F)
C7H4Cl2O
MFCD00000671
UN3265
Moisture sensitive
WHIHIKVIWVIIER-UHFFFAOYSA-N
3-chlorobenzoyl chloride
69252
97%
618-46-2
224°C to 225°C
Acidic
1.569
100 g
386256
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
ClC(=O)C1=CC=CC(Cl)=C1
175.01
175.01

Chemical Identifiers

618-46-2
175.01
WHIHIKVIWVIIER-UHFFFAOYSA-N
69252
ClC(=O)C1=CC=CC(Cl)=C1
C7H4Cl2O
MFCD00000671
m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
3-chlorobenzoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-552-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only