missing translation for 'onlineSavingsMsg'
Learn More

(3-Chlorobenzoyl)acetonitrile 97.0+%, TCI America™

Catalog No. p-7125541
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25811G 1 g
C25815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25811G Supplier TCI America Supplier No. C25811G
Only null left

Chemical Identifiers

CAS 21667-62-9
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.60
MDL Number MFCD00067891
InChI Key IUDFNNHFARLIPF-UHFFFAOYSA-N
Synonym 3-chlorobenzoylacetonitrile, 3-3-chlorophenyl-3-oxopropanenitrile, 3-chlorophenacylcyanide, 3-chlorobenzoyl acetonitrile, 3-3-chlorophenyl-3-oxo-propanenitrile, beta-ketonitrile 5b, pubchem12046, m-chlorophenacyl cyanide, acmc-1cqmb, maybridge1_004670
PubChem CID 140855
IUPAC Name 3-(3-chlorophenyl)-3-oxopropanenitrile
SMILES ClC1=CC=CC(=C1)C(=O)CC#N

Specifications

Melting Point 82°C
Quantity 1 g
Formula Weight 179.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (3-Chlorobenzoyl)acetonitrile
TSCA No
Recommended Storage Refrigerator