missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzotrifluoride 90.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 139°C |
| Quantity | 500 g |
| UN Number | 2234 |
| Formula Weight | 180.55 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Chlorobenzotrifluoride |
Safety and Handling
| EINECSNumber | (3)-0053 |
| RTECSNumber | XS9142000 |
| TSCA | Yes |