missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzophenone 98.0+%, TCI America™

Catalog No. p-7125161
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C177525G 25 g
C1775500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C177525G Supplier TCI America Supplier No. C177525G
Only null left

Chemical Identifiers

CAS 1016-78-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00009816
InChI Key CPLWKNRPZVNELG-UHFFFAOYSA-N
Synonym 3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
PubChem CID 66098
IUPAC Name (3-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl

Specifications

Melting Point 83°C
Color White
Boiling Point 332°C
Quantity 25 g
Formula Weight 216.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorobenzophenone
EINECSNumber (4)-0127
TSCA No