missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzonitrile, 99%, Thermo Scientific™

Chemical Identifiers

CAS 766-84-7
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001798
InChI Key WBUOVKBZJOIOAE-UHFFFAOYSA-N
Synonym m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
PubChem CID 13015
IUPAC Name 3-chlorobenzonitrile
SMILES ClC1=CC=CC(=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC108680100
SDS View Documents
Thermo Scientific Chemicals
108680100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC108680500
SDS View Documents
Thermo Scientific Chemicals
108680500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

766-84-7
137.57
WBUOVKBZJOIOAE-UHFFFAOYSA-N
13015
ClC1=CC=CC(=C1)C#N
C7H4ClN
MFCD00001798
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
3-chlorobenzonitrile

Specifications

766-84-7
100.0
Authentic
Glass bottle
ClC6H4CN
10g
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
WBUOVKBZJOIOAE-UHFFFAOYSA-N
3-chlorobenzonitrile
13015
99%
98.5
94°C (11.0mmHg)
98.5% min. (GC)
C7H4ClN
MFCD00001798
09, 339
Solubility in water: insoluble
ClC1=CC=CC(=C1)C#N
137.57
137.57
3-Chlorobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Warning

EINECSNumber : 212-172-6

SDS
Documents

Documents

RUO – Research Use Only