missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenethiol 97.0+%, TCI America™

Catalog No. p-7125071
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15825G 5 g
C158225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15825G Supplier TCI America Supplier No. C15825G
Only null left

Chemical Identifiers

CAS 2037-31-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
MDL Number MFCD00004839
InChI Key CQJDYPZUDYXHLM-UHFFFAOYSA-N
Synonym 3-chlorothiophenol, m-chlorothiophenol, benzenethiol, 3-chloro, 3-chlorobenzene-1-thiol, m-chlorobenzenethiol, benzenethiol, m-chloro, 3-chloro thiophenol, 3-chloro-benzenethiol, 3-chloromercaptobenzene, m-chlorophenyl mercaptan
PubChem CID 16257
IUPAC Name 3-chlorobenzene-1-thiol
SMILES SC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 1760
Formula Weight 144.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzenethiol