missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzamide 98.0+%, TCI America™
Supplier: TCI America C056225G
Specifications
| 3-Chlorobenzamide | |
| 618-48-4 | |
| 25 g | |
| C7H6ClNO | |
| m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce | |
| NC(=O)C1=CC=CC(Cl)=C1 | |
| 155.58 | |
| CHEBI:10587 | |
| ≥98.0% (N) |
| 134°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00014799 | |
| MJTGQALMWUUPQM-UHFFFAOYSA-N | |
| 3-chlorobenzamide | |
| 69254 | |
| 155.58 |
Chemical Identifiers
| 618-48-4 | |
| 155.58 | |
| MJTGQALMWUUPQM-UHFFFAOYSA-N | |
| 69254 | |
| 3-chlorobenzamide |
| C7H6ClNO | |
| MFCD00014799 | |
| m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce | |
| CHEBI:10587 | |
| NC(=O)C1=CC=CC(Cl)=C1 |
Safety and Handling
EINECSNumber : (3)-1430
RTECSNumber : CV2443774
TSCA : No