missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzamide 98.0+%, TCI America™

Catalog No. p-7124499
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
C056225G 25 g
C0562250G 250 g
2 options

Catalog No. C056225G

Supplier: TCI America C056225G

Only null left

Chemical Identifiers

CAS 618-48-4
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00014799
InChI Key MJTGQALMWUUPQM-UHFFFAOYSA-N
Synonym m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce
PubChem CID 69254
ChEBI CHEBI:10587
IUPAC Name 3-chlorobenzamide
SMILES NC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 155.58
Percent Purity ≥98.0% (N)
Chemical Name or Material 3-Chlorobenzamide
EINECSNumber (3)-1430
RTECSNumber CV2443774
TSCA No