missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzaldehyde 98.0+%, TCI America™
Supplier: TCI America C0124100G
Specifications
| 3-Chlorobenzaldehyde | |
| Yellow | |
| C7H5ClO | |
| 100 g | |
| SRWILAKSARHZPR-UHFFFAOYSA-N | |
| 3-chlorobenzaldehyde | |
| 11477 | |
| ≥98.0% (GC) |
| 587-04-2 | |
| 214°C | |
| MFCD00003350 | |
| m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
| C1=CC(=CC(=C1)Cl)C=O | |
| 140.566 | |
| 140.57 | |
| Liquid |
Chemical Identifiers
| 587-04-2 | |
| 140.566 | |
| SRWILAKSARHZPR-UHFFFAOYSA-N | |
| 11477 | |
| C1=CC(=CC(=C1)Cl)C=O |
| C7H5ClO | |
| MFCD00003350 | |
| m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde | |
| 3-chlorobenzaldehyde |
Safety and Handling
EINECSNumber : (3)-1162
TSCA : Yes