missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzaldehyde 98.0+%, TCI America™

Catalog No. C0124500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0124500G 500 g
C012425G 25 g
C0124100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0124500G Supplier TCI America Supplier No. C0124500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 587-04-2
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003350
InChI Key SRWILAKSARHZPR-UHFFFAOYSA-N
Synonym m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde
PubChem CID 11477
IUPAC Name 3-chlorobenzaldehyde
SMILES C1=CC(=CC(=C1)Cl)C=O

Specifications

Color Yellow
Boiling Point 214°C
Quantity 500 g
Formula Weight 140.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzaldehyde
EINECSNumber (3)-1162
TSCA Yes