missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzaldehyde, 99%

Catalog No. AC108611000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108611000 100 mL Glass bottle
AC108610250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108611000 Supplier Thermo Scientific Chemicals Supplier No. 108611000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 587-04-2 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 587-04-2
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00003350
InChI Key SRWILAKSARHZPR-UHFFFAOYSA-N
Synonym m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde
PubChem CID 11477
IUPAC Name 3-chlorobenzaldehyde
SMILES C1=CC(=CC(=C1)Cl)C=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 17°C to 18°C
Color Colorless to Yellow
Density 1.2410g/mL
Boiling Point 213°C to 215°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5630 to 1.565
Linear Formula ClC6H4CHO
Quantity 100 mL
Beilstein 07, 234
Specific Gravity 1.241
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 140.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-chlorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-596-9

RUO – Research Use Only