missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzaldehyde, 97%

CAS: 587-04-2 | C7H5ClO | 140.566 g/mol

Supplier:  Thermo Scientific Chemicals A1325422

 View more versions of this product

Catalog No. AAA1325422


Only null left
Explore available promotions

Description

Description

3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.

Solubility
It is soluble in alcohol, ether, and acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong bases, Strong oxidizing agents, Strong reducing agents.
Specifications

Specifications

3-Chlorobenzaldehyde
16°C to 18°C
213°C to 214°C
Characteristic
1.564
100 g
Air Sensitive
It is soluble in alcohol,ether,and acetone. Insoluble in water.
C1=CC(=CC(=C1)Cl)C=O
140.566
140.57
587-04-2
1.239
88°C (190°F)
C7H5ClO
MFCD00003350
507098
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde
SRWILAKSARHZPR-UHFFFAOYSA-N
3-chlorobenzaldehyde
11477
97%

Chemical Identifiers

587-04-2
140.566
SRWILAKSARHZPR-UHFFFAOYSA-N
11477
C1=CC(=CC(=C1)Cl)C=O
C7H5ClO
MFCD00003350
m-chlorobenzaldehyde, benzaldehyde, 3-chloro, mcad, meta-chlorobenzaldehyde, benzaldehyde, m-chloro, 3-chloro benzaldehyde, 3-chloro-benzaldehyde, benzaldehyde, 3-chloro-9ci, ccris 8613, 3-chiorobenzaldehyde
3-chlorobenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-596-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only