missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroanisole 98.0+%, TCI America™

Catalog No. C114825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C114825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C114825G Supplier TCI America Supplier No. C114825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2845-89-8
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000591
InChI Key YUKILTJWFRTXGB-UHFFFAOYSA-N
Synonym 3-chloroanisole, m-chloroanisole, benzene, 1-chloro-3-methoxy, anisole, m-chloro, 3-chloroanisol, meta-chloroanisole, 3-anisolyl chloride, pubchem3615, 3-chloromethoxybenzene, m-methoxyphenyl chloride
PubChem CID 17833
IUPAC Name 1-chloro-3-methoxybenzene
SMILES COC1=CC(=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 193°C
Quantity 25 g
Formula Weight 142.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloroanisole