missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroacetylacetone 95.0+%, TCI America™

Catalog No. C127725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C127725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C127725G Supplier TCI America Supplier No. C127725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1694-29-7
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.559
MDL Number MFCD00009651
InChI Key VLRGXXKFHVJQOL-UHFFFAOYSA-N
Synonym 3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
PubChem CID 74328
IUPAC Name 3-chloropentane-2,4-dione
SMILES CC(=O)C(C(=O)C)Cl

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 g
UN Number 1224
Formula Weight 134.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloroacetylacetone