accessibility menu, dialog, popup

UserName

CH3OClCH3O

CAS RN 1694-29-7

IUPAC Name: 3-chloropentane-2,4-dione
Synonyms: 3-chloropentane-2,4-dione
Molecular Weight (g/mol): 134.56
Molecular Formula: C5H7ClO2
InChi Key: VLRGXXKFHVJQOL-UHFFFAOYSA-N
SMILES: CC(=O)C(Cl)C(C)=O
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)