missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-p-cresol 98.0+%, TCI America™

Catalog No. p-7125929
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33335G 5 g
C333325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33335G Supplier TCI America Supplier No. C33335G
Only null left

Chemical Identifiers

CAS 615-62-3
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060319
InChI Key VQZRLBWPEHFGCD-UHFFFAOYSA-N
PubChem CID 14853
IUPAC Name 3-chloro-4-methylphenol
SMILES CC1=CC=C(O)C=C1Cl

Specifications

Melting Point 53°C
Boiling Point 228°C
Quantity 5 g
UN Number 3437
Synonym phenol, 3-chloro-4-methyl, 3-chloro-p-cresol, p-cresol, 3-chloro, 3-chloro-4-methyl-phenol, acmc-20aibl, pubchem3671, 2-chloro-4-hydroxytoluene, phenol,3-chloro-4-methyl, p-cresol, 3-chloro-8ci, 3-chloro-4-methylphenol
Formula Weight 142.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-p-cresol
EINECSNumber (3)-0900
TSCA Yes