missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-o-xylene, 97%

Catalog No. AAB2150522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2150522 100 g
AAB2150514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2150522 Supplier Thermo Scientific Chemicals Supplier No. B2150522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 608-23-1 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608-23-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00060689
InChI Key NVLHGZIXTRYOKT-UHFFFAOYSA-N
Synonym 3-chloro-o-xylene, 2,3-dimethylchlorobenzene, benzene, chlorodimethyl, 3-chloro-1,2-dimethylbenzene, 3-chloro-ortho-xylene, benzene, 1-chloro-2,3-dimethyl, xylyl chloride, chloro o-xylene, mono-chlorxylol, chloro-ortho-xylene
PubChem CID 32884
IUPAC Name 1-chloro-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)Cl)C

Specifications

Quantity 100 g
Formula Weight 140.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-o-xylene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only