missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-N-methylaniline 97.0+%, TCI America™
$213.69 - $852.85
Chemical Identifiers
| CAS | 7006-52-2 |
|---|---|
| Molecular Formula | C7H8ClN |
| Molecular Weight (g/mol) | 141.60 |
| MDL Number | MFCD00052014 |
| InChI Key | WFGYSQDPURFIFL-UHFFFAOYSA-N |
| Synonym | n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline |
| PubChem CID | 138900 |
| IUPAC Name | 3-chloro-N-methylaniline |
| SMILES | CNC1=CC=CC(Cl)=C1 |
Chemical Identifiers
| 7006-52-2 | |
| 141.60 | |
| WFGYSQDPURFIFL-UHFFFAOYSA-N | |
| 138900 | |
| CNC1=CC=CC(Cl)=C1 |
| C7H8ClN | |
| MFCD00052014 | |
| n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline | |
| 3-chloro-N-methylaniline |
Specifications
| 7006-52-2 | |
| 235°C | |
| MFCD00052014 | |
| n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline | |
| CNC1=CC=CC(Cl)=C1 | |
| 141.60 | |
| 141.60 | |
| Liquid |
| Yellow | |
| C7H8ClN | |
| 5 g | |
| WFGYSQDPURFIFL-UHFFFAOYSA-N | |
| 3-chloro-N-methylaniline | |
| 138900 | |
| ≥97.0% (GC,T) | |
| 3-Chloro-N-methylaniline |
Safety and Handling
TSCA : No