missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-L-phenylalanine, 95%

Catalog No. AAH5201803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5201803 1 g
AAH5201806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5201803 Supplier Thermo Scientific Chemicals Supplier No. H5201803

CAS: 80126-51-8 | C9H10ClNO2 | 199.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80126-51-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD01075124
InChI Key JJDJLFDGCUYZMN-QMMMGPOBSA-N
Synonym 3-chloro-l-phenylalanine, l-3-chlorophenylalanine, s-2-amino-3-3-chlorophenyl propanoic acid, h-phe 3-cl-oh, 2s-2-amino-3-3-chlorophenyl propanoic acid, 3-chlorophenylalanine, l-phe 3-cl-oh, 3-chloro-l-phenylalanine hydrochloride, d-3-chlorophenylalanine, l-3-chlorophe
PubChem CID 2761492
ChEBI CHEBI:49597
IUPAC Name (2S)-2-amino-3-(3-chlorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(Cl)=C1)C(O)=O

Specifications

Quantity 1 g
Formula Weight 199.64
Percent Purity 95%
Chemical Name or Material 3-Chloro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only