accessibility menu, dialog, popup

UserName

H2N(S)ClHOO

CAS RN 80126-51-8

IUPAC Name: (2S)-2-amino-3-(3-chlorophenyl)propanoic acid
Synonyms: C9H10ClNO2
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: JJDJLFDGCUYZMN-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=CC(Cl)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)