missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-L-alanine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7125089
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16121G Supplier TCI America Supplier No. C16121G

Chemical Identifiers

CAS 51887-89-9
Molecular Formula C3H7Cl2NO2
Molecular Weight (g/mol) 159.994
MDL Number MFCD00070398
InChI Key IENJPSDBNBGIEL-DKWTVANSSA-N
Synonym (R)-2-Amino-3-chloropropionic Acid Hydrochloride, beta-Chloro-L-alanine Hydrochloride, H-Ala(3-Cl)-OH.HCl
PubChem CID 11957493
IUPAC Name (2R)-2-amino-3-chloropropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)Cl.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 159.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-L-alanine Hydrochloride