missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-DL-mandelic Acid 97.0+%, TCI America™
$86.64 - $286.85
Chemical Identifiers
| CAS | 16273-37-3 |
|---|---|
| Molecular Formula | C8H7ClO3 |
| Molecular Weight (g/mol) | 186.591 |
| InChI Key | SAMVPMGKGGLIPF-UHFFFAOYSA-N |
| Synonym | 3-chloromandelic acid, 2-3-chlorophenyl-2-hydroxyacetic acid, 3-chlorophenylglycolic acid, 3-chlorophenyl hydroxy acetic acid, 3-chloromandelicacid, benzeneacetic acid,3-chloro-a-hydroxy-, ar, maybridge3_000705, 3-chloro-dl-mandelic acid, acmc-1c3h4, racemic 3-chloromandelic acid |
| PubChem CID | 85981 |
| IUPAC Name | 2-(3-chlorophenyl)-2-hydroxyacetic acid |
| SMILES | C1=CC(=CC(=C1)Cl)C(C(=O)O)O |
Chemical Identifiers
| 16273-37-3 | |
| 186.591 | |
| 3-chloromandelic acid, 2-3-chlorophenyl-2-hydroxyacetic acid, 3-chlorophenylglycolic acid, 3-chlorophenyl hydroxy acetic acid, 3-chloromandelicacid, benzeneacetic acid,3-chloro-a-hydroxy-, ar, maybridge3_000705, 3-chloro-dl-mandelic acid, acmc-1c3h4, racemic 3-chloromandelic acid | |
| 2-(3-chlorophenyl)-2-hydroxyacetic acid |
| C8H7ClO3 | |
| SAMVPMGKGGLIPF-UHFFFAOYSA-N | |
| 85981 | |
| C1=CC(=CC(=C1)Cl)C(C(=O)O)O |
Specifications
| 115°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| 3-chloromandelic acid, 2-3-chlorophenyl-2-hydroxyacetic acid, 3-chlorophenylglycolic acid, 3-chlorophenyl hydroxy acetic acid, 3-chloromandelicacid, benzeneacetic acid,3-chloro-a-hydroxy-, ar, maybridge3_000705, 3-chloro-dl-mandelic acid, acmc-1c3h4, racemic 3-chloromandelic acid | |
| C1=CC(=CC(=C1)Cl)C(C(=O)O)O | |
| 186.591 | |
| 186.59 | |
| 3-Chloro-DL-mandelic Acid |
| 16273-37-3 | |
| 5 g | |
| C8H7ClO3 | |
| SAMVPMGKGGLIPF-UHFFFAOYSA-N | |
| 2-(3-chlorophenyl)-2-hydroxyacetic acid | |
| 85981 | |
| ≥97.0% (GC,T) |
Safety and Handling
TSCA : No