missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-DL-mandelic Acid 97.0+%, TCI America™

Supplier:  TCI America C200925G

 View more versions of this product

Catalog No. C200925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Chloro-DL-mandelic Acid
16273-37-3
25 g
C8H7ClO3
SAMVPMGKGGLIPF-UHFFFAOYSA-N
2-(3-chlorophenyl)-2-hydroxyacetic acid
85981
≥97.0% (GC,T)
115°C
White-Yellow
Crystal-Powder at 20°C
3-chloromandelic acid, 2-3-chlorophenyl-2-hydroxyacetic acid, 3-chlorophenylglycolic acid, 3-chlorophenyl hydroxy acetic acid, 3-chloromandelicacid, benzeneacetic acid,3-chloro-a-hydroxy-, ar, maybridge3_000705, 3-chloro-dl-mandelic acid, acmc-1c3h4, racemic 3-chloromandelic acid
C1=CC(=CC(=C1)Cl)C(C(=O)O)O
186.591
186.59

Chemical Identifiers

16273-37-3
186.591
3-chloromandelic acid, 2-3-chlorophenyl-2-hydroxyacetic acid, 3-chlorophenylglycolic acid, 3-chlorophenyl hydroxy acetic acid, 3-chloromandelicacid, benzeneacetic acid,3-chloro-a-hydroxy-, ar, maybridge3_000705, 3-chloro-dl-mandelic acid, acmc-1c3h4, racemic 3-chloromandelic acid
2-(3-chlorophenyl)-2-hydroxyacetic acid
C8H7ClO3
SAMVPMGKGGLIPF-UHFFFAOYSA-N
85981
C1=CC(=CC(=C1)Cl)C(C(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions