missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methylpyridazine 98.0+%, TCI America™

Catalog No. p-7125378
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22745G 5 g
C227425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22745G Supplier TCI America Supplier No. C22745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1121-79-5
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00052905
InChI Key PRORLQAJNJMGAR-UHFFFAOYSA-N
Synonym 3-chloro-6-methyl-pyridazine, 3-chloro-6-methyl pyridazine, pyridazine, 3-chloro-6-methyl, 6-chloro-3-methylpyridazine, pubchem10467, 3-methyl-6-chloropyridazine, 6-methyl-3-chloropyridazine, 3-chloro-6-methyl-1,2-diazine, 3-chloro-6-methylpyridazine
PubChem CID 227254
IUPAC Name 3-chloro-6-methylpyridazine
SMILES CC1=NN=C(C=C1)Cl

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-6-methylpyridazine