CAS RN 1121-79-5
IUPAC Name:
3-chloro-6-methylpyridazine
Synonyms:
3-chloro-6-methylpyridazine
Molecular Weight (g/mol):
128.56
Molecular Formula:
C5H5ClN2
InChi Key:
PRORLQAJNJMGAR-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Cl)N=N1
1
–
1
of
1
results
1
3-Chloro-6-methylpyridazine 98.0+%, TCI America™
CAS: 1121-79-5 Molecular Formula: C5H5ClN2 Molecular Weight (g/mol): 128.559 MDL Number: MFCD00052905 InChI Key: PRORLQAJNJMGAR-UHFFFAOYSA-N Synonym: 3-chloro-6-methyl-pyridazine,3-chloro-6-methyl pyridazine,pyridazine, 3-chloro-6-methyl,6-chloro-3-methylpyridazine,pubchem10467,3-methyl-6-chloropyridazine,6-methyl-3-chloropyridazine,3-chloro-6-methyl-1,2-diazine,3-chloro-6-methylpyridazine PubChem CID: 227254 IUPAC Name: 3-chloro-6-methylpyridazine SMILES: CC1=NN=C(C=C1)Cl