Learn More
3-Chloro-6-iodopyridazine, 95%
CAS: 135034-10-5 | C4H2ClIN2 | 240.43 g/mol
$111.74 - $474.86
Chemical Identifiers
| CAS | 135034-10-5 |
|---|---|
| Molecular Formula | C4H2ClIN2 |
| Molecular Weight (g/mol) | 240.43 |
| MDL Number | MFCD08275187 |
| InChI Key | PNEPCDPKMXJYIQ-UHFFFAOYSA-N |
| Synonym | 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine |
| PubChem CID | 15418839 |
| IUPAC Name | 3-chloro-6-iodopyridazine |
| SMILES | ClC1=NN=C(I)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6387903
|
Thermo Scientific Chemicals
H6387903 |
1 g |
Each for $111.74
|
|
|||||
|
AAH6387906
|
Thermo Scientific Chemicals
H6387906 |
5 g |
Each for $474.86
|
|
|||||
Description
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
Chemical Identifiers
| 135034-10-5 | |
| 240.43 | |
| PNEPCDPKMXJYIQ-UHFFFAOYSA-N | |
| 15418839 | |
| ClC1=NN=C(I)C=C1 |
| C4H2ClIN2 | |
| MFCD08275187 | |
| 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine | |
| 3-chloro-6-iodopyridazine |
Specifications
| 135034-10-5 | |
| C4H2ClIN2 | |
| 1 g | |
| 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine | |
| PNEPCDPKMXJYIQ-UHFFFAOYSA-N | |
| 3-chloro-6-iodopyridazine | |
| 15418839 | |
| 97% | |
| 3-Chloro-6-iodopyridazine |
| 110°C to 120°C | |
| MFCD08275187 | |
| Light sensitive | |
| Slightly soluble in water. | |
| ClC1=NN=C(I)C=C1 | |
| 240.43 | |
| 240.43 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only