missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-iodopyridazine, 95%

Catalog No. AAH6387903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6387903 1 g
AAH6387906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6387903 Supplier Thermo Scientific Chemicals Supplier No. H6387903

CAS: 135034-10-5 | C4H2ClIN2 | 240.43 g/mol

Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reacant in Suzuki-Miyaura coupling reaction. Reactant in the synthesis of 6-Chloropyridazine-3-carbonitrile.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.

Chemical Identifiers

CAS 135034-10-5
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.43
MDL Number MFCD08275187
InChI Key PNEPCDPKMXJYIQ-UHFFFAOYSA-N
Synonym 3-chloro-6-iodo-pyridazine, pyridazine, 3-chloro-6-iodo, 3-chloro-6-pyrazine, acmc-1cg9b, ksc169s6d, 3-chloranyl-6-iodanyl-pyridazine, 6-chloro-3-iodopyridazine, 3-chloro-6-iodo-1,2-diazine
PubChem CID 15418839
IUPAC Name 3-chloro-6-iodopyridazine
SMILES ClC1=NN=C(I)C=C1

Specifications

Melting Point 110°C to 120°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 240.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Chloro-6-iodopyridazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only