missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7032695
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3425803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3425803 Supplier Thermo Scientific Chemicals Supplier No. H3425803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80194-68-9 | Molecular Formula: C7H3ClF3NO2 | Molecular Weight: 225.551 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80194-68-9
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD00277482
InChI Key HXRMCZBDTDCCOP-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
PubChem CID 2821908
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Formula Weight 225.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only